4-Fluoro-2-formylphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-formylbenzeneboronic acid, (4-Fluoro-2-formylphenyl)boronic acid

A177588-1G A177588-250MG A177588-25G A177588-5G
76974-306EA 7 USD
76974-306 76974-308 76974-310 76974-312
4-Fluoro-2-formylphenylboronic acid 95%
4-Fluoro-2-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₃
MW: 167.93 g/mol
MDL Number: MFCD10697426
CAS Number: 825644-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR