4-Chloro-2-fluoroanisole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-2-fluoro-1-methoxybenzene
A185766-1G
A185766-25G
A185766-5G
76974-618EA
27.59
USD
76974-618
76974-620
76974-622
4-Chloro-2-fluoroanisole 97%
4-Chloro-2-fluoroanisole
Precaución: for Research Use Only
Formula:
C₇H₆ClFO MW: 160.58 g/mol |
MDL Number:
MFCD00672964 CAS Number: 452-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...