6-(Methylamino)pyrimidin-4-ol 97%

Supplier: Ambeed
Warning

A504417-100MG A504417-1G A504417-250MG
76979-908EA 118.83 USD
76979-908 76979-910 76979-912
6-(Methylamino)pyrimidin-4-ol 97%
6-(Methylamino)pyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.13 g/mol
MDL Number: MFCD18762029
CAS Number: 1122-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR