2-Bromo-6-methyl-4-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A611276-100MG A611276-1G A611276-250MG A611276-5G
76980-556EA 7 USD
76980-556 76980-558 76980-560 76980-562
2-Bromo-6-methyl-4-(trifluoromethyl)pyridine 95%
2-Bromo-6-methyl-4-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrF₃N
MDL Number: MFCD07368051
CAS Number: 451459-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR