1,2,3,4-Tetrahydro-2,6-naphthyridine 98%

Supplier: Ambeed
Warning

A700574-250MG A700574-100MG
76981-096EA 273.72 USD
76981-096 77673-918
1,2,3,4-Tetrahydro-2,6-naphthyridine 98%
1,2,3,4-Tetrahydro-2,6-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂
MW: 134.1784 g/mol
MDL Number: MFCD08752595
CAS Number: 31786-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR