(1R,3S)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1R,3S)-N-BOC-3-aminocyclohexanecarboxylic acid, (1R,3S)-3-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid, (1R,3S)-3-(Boc-amino)cyclohexanecarboxylic Acid

A116037-100MG A116037-1G A116037-250MG A116037-5G
76971-170EA 27 USD
76971-170 76971-172 76971-174 77670-628
(1R,3S)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 95%
(1R,3S)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD04973107
CAS Number: 222530-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR