5,7-Dichlorobenzo[d]thiazol-2-amine 97%

Supplier: Ambeed
Warning

A546836-100MG A546836-1G A546836-250MG A546836-5G
76980-218EA 18.04 USD
76980-218 76980-220 76980-222 76980-224
5,7-Dichlorobenzo[d]thiazol-2-amine 97%
5,7-Dichlorobenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂S
MW: 219.09 g/mol
MDL Number: MFCD10693482
CAS Number: 158465-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR