2-(Phenylsulfonyl)pyridine 95%

Supplier: Ambeed
Warning

A110136-10G A110136-1G A110136-250MG A110136-25G A110136-5G
76970-684EA 89.12 USD
76970-684 76970-686 76970-688 76970-690 76970-692
2-(Phenylsulfonyl)pyridine 95%
2-(Phenylsulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂S
MW: 219.2600 g/mol
MDL Number: MFCD09999263
CAS Number: 24244-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR