1-Boc-2-azetidinemethanol 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(hydroxymethyl)azetidine-1-carboxylate

A604151-10G A604151-25G A604151-1G
76980-522EA 152.77 USD
76980-522 76980-524 77673-628
1-Boc-2-azetidinemethanol 98%
1-Boc-2-azetidinemethanol

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
MDL Number: MFCD03790936
CAS Number: 174346-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR