4-Fluoro-3-methoxybenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: 5-Aminomethyl-2-fluoroanisole, 5-(Aminomethyl)-2-fluoroanisole

A915643-1G A915643-250MG A915643-5G
76981-944EA 76.39 USD
76981-944 76981-946 76981-948
4-Fluoro-3-methoxybenzylamine 98%
4-Fluoro-3-methoxybenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀FNO
MW: 155.1700 g/mol
MDL Number: MFCD04116362
CAS Number: 508177-67-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR