2-Chloro-6-(trifluoromethyl)isonicotinonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-(trifluoromethyl)pyridine-4-carbonitrile

A278448-1G A278448-250MG
76977-340EA 99.73 USD
76977-340 76977-342
2-Chloro-6-(trifluoromethyl)isonicotinonitrile 95%
2-Chloro-6-(trifluoromethyl)isonicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₃N₂
MW: 206.552 g/mol
MDL Number: MFCD13190042
CAS Number: 1196155-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR