4-Amino-3-chloro-5-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-5-methylpyridin-4-amine

A148523-1G A148523-250MG
76972-986EA 136.86 USD
76972-986 76972-988
4-Amino-3-chloro-5-methylpyridine 95%
4-Amino-3-chloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.5862 g/mol
MDL Number: MFCD08277270
CAS Number: 97944-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR