Benzo[d]oxazol-2-ylmethanol 96%

Supplier: Ambeed
Warning

A141850-100MG A141850-1G A141850-250MG A141850-5G
76972-628EA 16.98 USD
76972-628 76972-630 76972-632 76972-634
Benzo[d]oxazol-2-ylmethanol 96%
Benzo[d]oxazol-2-ylmethanol

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.1467 g/mol
MDL Number: MFCD07368333
CAS Number: 77186-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR