7-Bromo-2-chlorothieno[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A146211-1G A146211-5G A146211-250MG A146211-100MG
76972-852EA 163 USD
76972-852 76972-854 77611-348 77881-140
7-Bromo-2-chlorothieno[3,2-d]pyrimidine 95%
7-Bromo-2-chlorothieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂S
MDL Number: MFCD13193398
CAS Number: 1152475-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR