N-Carbobenzoxy-DL-methionine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cbz-DL-Met-OH, N-Cbz-DL-methionine, Z-DL-Met-OH
A186990-100G
A186990-1G
A186990-25G
A186990-5G
76974-692EA
362
USD
76974-692
76974-694
77671-644
77671-646
N-Carbobenzoxy-DL-methionine 98%
N-Carbobenzoxy-DL-methionine
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₄S MW: 283.34 g/mol |
MDL Number:
MFCD00065123 CAS Number: 4434-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...