Methyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methyl-4-methoxycarbonylphenylboronic acid, pinacol ester
A376356-1G
A376356-250MG
A376356-25G
A376356-5G
76978-780EA
27.59
USD
76978-780
76978-782
76978-784
76978-786
Methyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98%
Methyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₅H₂₁BO₄ MW: 276.13 g/mol |
MDL Number:
MFCD08669595 CAS Number: 473596-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...