(4-Chloro-2-ethylphenyl)amine 95%

Supplier: Ambeed
Warning

A229502-25G A229502-5G A229502-1G
76976-094EA 663.07 USD
76976-094 76976-096 77612-234
(4-Chloro-2-ethylphenyl)amine 95%
(4-Chloro-2-ethylphenyl)amine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
MW: 155.63 g/mol
MDL Number: MFCD01670148
CAS Number: 30273-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR