(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-4,4'-Bis(bis(3,5-dimethylphenyl)phosphino)-2,2',6,6'-tetramethoxy-3,3'-bipyridine, CTH-(R)-Xylyl-P-PHOS

A575537-100MG A575537-1G A575537-250MG A575537-50MG
76980-360EA 111 USD
76980-360 76980-362 76980-364 76980-366
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine 98%
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine

Precaución: for Research Use Only

Formula: C₄₆H₅₀N₂O₄P₂
MDL Number: MFCD04974235
CAS Number: 442905-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR