Methyl-3-fluoro-4-methylbenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-fluoro-4-methylbenzoate
A768614-100G
A768614-10G
A768614-1G
A768614-5G
76981-482EA
309.79
USD
76981-482
76981-484
76981-486
76981-488
Methyl-3-fluoro-4-methylbenzoate 98%
Methyl 3-fluoro-4-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₉FO₂ MW: 168.1650 g/mol |
MDL Number:
MFCD06203786 CAS Number: 87808-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...