4-Bromo-3-nitro-1H-pyrazole 97%

Supplier: Ambeed
Warning

A140083-100MG A140083-1G A140083-250MG A140083-5G A140083-25G
76972-556EA 9 USD
76972-556 76972-558 76972-560 76972-562 77881-097
4-Bromo-3-nitro-1H-pyrazole 97%
4-Bromo-3-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃H₂BrN₃O₂
MDL Number: MFCD01114992
CAS Number: 89717-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR