Tizoxanide 98%

Supplier: Ambeed
Warning

A108453-10MG A108453-1MG A108453-5MG
76970-562EA 57 USD
76970-562 76970-564 76970-566
Tizoxanide 98%
Tizoxanide

Precaución: for Research Use Only

Formula: C₁₀H₇N₃O₄S
MDL Number: MFCD07484970
CAS Number: 173903-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR