3-Hydroxy-1-methylazetidine 96%

Supplier: Ambeed
Warning

Synonyms: 1-Methylazetidin-3-ol, 1-Methyl-3-azetidinol

A286021-100MG A286021-1G A286021-250MG
76977-506EA 94.43 USD
76977-506 76977-508 76977-510
3-Hydroxy-1-methylazetidine 96%
3-Hydroxy-1-methylazetidine

Precaución: for Research Use Only

Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD10565802
CAS Number: 111043-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR