2-Amino-4-(trifluoromethyl)pyrimidine-5-carbaldehyde 98%

Supplier: Ambeed
Warning

A317515-100MG A317515-1G A317515-50MG
76978-006EA 160.2 USD
76978-006 76978-008 76978-010
2-Amino-4-(trifluoromethyl)pyrimidine-5-carbaldehyde 98%
2-Amino-4-(trifluoromethyl)pyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₄F₃N₃O
MW: 191.11 g/mol
MDL Number: MFCD13193531
CAS Number: 863774-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR