tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(2-hydroxyethyl)azetidine-1-carboxylate

A222633-100MG A222633-1G A222633-250MG A222633-5G A222633-25G
76975-870EA 41.38 USD
76975-870 76975-872 76975-874 77593-442 77612-158
tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate 96%
tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD17015912
CAS Number: 152537-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR