5-Chloro-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carbaldehyde 98%

Supplier: Ambeed
Warning

A124308-100MG A124308-10G A124308-1G A124308-250MG A124308-25G A124308-5G
76971-756EA 21.22 USD
76971-756 76971-758 76971-760 76971-762 76971-764 76971-766
5-Chloro-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carbaldehyde 98%
5-Chloro-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₅ClF₂N₂O
MW: 194.567 g/mol
MDL Number: MFCD12827692
CAS Number: 660845-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR