2-Fluoro-6-iodonitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Fluoro-3-iodo-2-nitrobenzene

A135207-100MG A135207-10G A135207-1G A135207-250MG A135207-5G
76972-306EA 9.55 USD
76972-306 76972-308 76972-310 76972-312 76972-314
2-Fluoro-6-iodonitrobenzene 97%
2-Fluoro-6-iodonitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃FINO₂
MW: 266.996 g/mol
MDL Number: MFCD07368790
CAS Number: 886762-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR