1,3,4-Oxadiazole 96%

Supplier: Ambeed
Warning

A829846-100MG A829846-250MG
76981-764EA 82.76 USD
76981-764 76981-766
1,3,4-Oxadiazole 96%
1,3,4-Oxadiazole

Precaución: for Research Use Only

Formula: C₂H₂N₂O
MW: 70.05 g/mol
MDL Number: MFCD00955535
CAS Number: 288-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR