2,6-Difluoro-4-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Difluoro-4-methoxynitrobenzene

A771470-1G A771470-5G
76981-502EA 20.16 USD
76981-502 76981-504
2,6-Difluoro-4-nitroanisole 98%
2,6-Difluoro-4-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₃
MW: 189.116 g/mol
MDL Number: MFCD06656243
CAS Number: 392-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR