6-Amino-9-(tetrahydropyran-2-yl)-9H-purine 97%

Supplier: Ambeed
Warning

Synonyms: 9-(Tetrahydro-2H-pyran-2-yl)-9H-purin-6-amine

A107076-100MG A107076-250MG A107076-5G
76970-478EA 26.53 USD
76970-478 76970-480 77670-530
6-Amino-9-(tetrahydropyran-2-yl)-9H-purine 97%
6-Amino-9-(tetrahydropyran-2-yl)-9H-purine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₅O
MW: 219.243 g/mol
MDL Number: MFCD00213333
CAS Number: 7306-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR