5-Methyl-1H-imidazo[4,5-b]pyridine 96%

Supplier: Ambeed
Warning

A190494-100MG A190494-1G A190494-250MG
76974-834EA 11 USD
76974-834 76974-836 76974-838
5-Methyl-1H-imidazo[4,5-b]pyridine 96%
5-Methyl-1H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MDL Number: MFCD07440139
CAS Number: 27582-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR