4-Amino-2-chloro-5-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-methylpyridin-4-amine

A173224-100G A173224-10G A173224-1G A173224-250MG A173224-25G A173224-5G
76974-118EA 126 USD
76974-118 76974-120 76974-122 76974-124 76974-126 76974-128
4-Amino-2-chloro-5-methylpyridine 95%
4-Amino-2-chloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MDL Number: MFCD11977435
CAS Number: 79055-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR