3,6-Dichloroimidazo[1,2-b]pyridazine 97%

Supplier: Ambeed
Warning

A155579-1G A155579-250MG
76973-300EA 80 USD
76973-300 76973-302
3,6-Dichloroimidazo[1,2-b]pyridazine 97%
3,6-Dichloroimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂N₃
MDL Number: MFCD11520887
CAS Number: 40972-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR