Ethyl 2-cyclopentylacetate 95%

Supplier: Ambeed
Warning

A444950-250MG A444950-5G
76979-422EA 57 USD
76979-422 76979-424
Ethyl 2-cyclopentylacetate 95%
Ethyl 2-cyclopentylacetate

Precaución: for Research Use Only

Formula: C₉H₁₆O₂
MDL Number: MFCD00019300
CAS Number: 18322-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR