2-Amino-6-nitrophenol 95%

Supplier: Ambeed
Warning

A213524-100MG A213524-250MG A213524-1G A213524-5G
76975-602EA 48 USD
76975-602 76975-604 77881-515 77881-516
2-Amino-6-nitrophenol 95%
2-Amino-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MDL Number: MFCD02751769
CAS Number: 603-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR