4-Methyl-2-nitro-1-(trifluoromethyl)benzene 95%

Supplier: Ambeed
Warning

A102566-10G A102566-1G A102566-250MG A102566-5G
76970-232EA 283 USD
76970-232 76970-234 76970-236 76970-238
4-Methyl-2-nitro-1-(trifluoromethyl)benzene 95%
4-Methyl-2-nitro-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MDL Number: MFCD16140172
CAS Number: 154057-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR