5-Methylpyridin-3-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline
A179821-100MG
A179821-10G
A179821-1G
A179821-250MG
A179821-5G
A179821-25G
76974-406EA
8
USD
76974-406
76974-408
76974-410
76974-412
77611-740
77671-542
5-Methylpyridin-3-ol 98%
5-Methylpyridin-3-ol
Precaución: for Research Use Only
Formula:
C₆H₇NO MW: 109.13 g/mol |
MDL Number:
MFCD00661297 CAS Number: 42732-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...