4,7,10-Trioxa-1,13-tridecanediamine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,13-Diamino-4,7,10-trioxatridecane, 3,3'-((Oxybis(ethane-2,1-diyl))bis(oxy))bis(propan-1-amine)
A221158-1000G
A221158-100G
A221158-10G
A221158-25G
A221158-500G
A221158-5G
76975-836EA
240.83
USD
76975-836
76975-838
76975-840
76975-842
76975-844
76975-846
4,7,10-Trioxa-1,13-tridecanediamine 95%
4,7,10-Trioxa-1,13-tridecanediamine
Precaución: for Research Use Only
Formula:
C₁₀H₂₄N₂O₃ MW: 220.3092 g/mol |
MDL Number:
MFCD00059850 CAS Number: 4246-51-9 UN: 2735 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...