2-Chloro-4,6-diamino-1,3,5-triazine 90%

Supplier: Ambeed
Warning

Synonyms: 2,4-Diamino-6-chloro-1,3,5-triazine, 6-Chloro-1,3,5-triazine-2,4-diamine

A159124-100G A159124-10G A159124-1G A159124-25G A159124-5G
76973-448EA 298.12 USD
76973-448 76973-450 76973-452 76973-454 76973-456
2-Chloro-4,6-diamino-1,3,5-triazine 90%
2-Chloro-4,6-diamino-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₃H₄ClN₅
MW: 145.55 g/mol
MDL Number: MFCD00006054
CAS Number: 3397-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR