(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one 99+%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydrobenzo[b][1,4]thiazepin-4(5H)-one

A156981-100G A156981-25G A156981-500G A156981-5G
76973-346EA 32.89 USD
76973-346 76973-348 76973-350 76973-352
(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one 99+%
(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₃S
MW: 301.36 g/mol
MDL Number: MFCD01321316
CAS Number: 42399-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR