3-Aminopyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-2-pyridinecarbonitrile

A524871-1G A524871-250MG A524871-25G A524871-5G
76980-032EA 61.54 USD
76980-032 76980-034 76980-036 76980-038
3-Aminopyridine-2-carbonitrile 98%
3-Aminopyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.1240 g/mol
MDL Number: MFCD02181031
CAS Number: 42242-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR