2-Bromo-4-nitro-6-(trifluoromethyl)aniline 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-3-bromo-5-nitrobenzotrifluoride
A364571-100MG
A364571-250MG
A364571-1G
76978-628EA
33
USD
76978-628
76978-630
77672-832
2-Bromo-4-nitro-6-(trifluoromethyl)aniline 95%
2-Bromo-4-nitro-6-(trifluoromethyl)aniline
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃N₂O₂ MW: 285.0200 g/mol |
MDL Number:
MFCD14560537 CAS Number: 400-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...