4-Iodo-3-methoxybenzenecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-3-methoxybenzoic acid

A212252-100MG A212252-1G A212252-250MG A212252-5G
76975-574EA 8.49 USD
76975-574 76975-576 76975-578 76975-580
4-Iodo-3-methoxybenzenecarboxylic acid 98%
4-Iodo-3-methoxybenzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₃
MW: 278.05 g/mol
MDL Number: MFCD09753732
CAS Number: 282087-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR