2,3,6-Tribromopyridine 98%

Supplier: Ambeed
Warning

A411085-100MG A411085-1G
76979-142EA 50.93 USD
76979-142 76979-144
2,3,6-Tribromopyridine 98%
2,3,6-Tribromopyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₃N
MDL Number: MFCD00234000
CAS Number: 2402-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR