(S)-(-)-4-Methoxy-α-methylbenzylamine 95%

Supplier: Ambeed
Danger

A498970-1G A498970-25G A498970-5G A498970-100G A498970-10G
76979-848EA 14 USD
76979-848 76979-850 76979-852 77882-235 77882-236
(S)-(-)-4-Methoxy-α-methylbenzylamine 95%
(S)-(-)-4-Methoxy-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MDL Number: MFCD00671660
CAS Number: 41851-59-6
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR