N-Methyl-L-isoleucine 95%

Supplier: Ambeed
Warning

A709671-1G A709671-250MG A709671-25G
76981-156EA 23 USD
76981-156 76981-158 77882-576
N-Methyl-L-isoleucine 95%
N-Methyl-L-isoleucine

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MDL Number: MFCD00037755
CAS Number: 4125-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR