6-Methoxybenzo[d]thiazol-2(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-2(3H)-benzothiazolone

A198021-1G A198021-5G
76975-144EA 47 USD
76975-144 76975-146
6-Methoxybenzo[d]thiazol-2(3H)-one 98%
6-Methoxybenzo[d]thiazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₈H₇NO₂S
MDL Number: MFCD09743851
CAS Number: 40925-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR