L-Tyrosine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr-OEt·HCl, H-Tyr-OEt.HCl

A133606-1G A133606-25G A133606-500G A133606-5G A133606-100G
76972-226EA 9 USD
76972-226 76972-228 76972-230 76972-232 77670-904
L-Tyrosine ethyl ester hydrochloride 98%
L-Tyrosine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MDL Number: MFCD00063047
CAS Number: 4089-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR