1,3,5-Tris(4-(tert-butyl)-3-hydroxy-2,6-dimethylbenzyl)-1,3,5-triazinane-2,4,6-trione 97%

Supplier: Ambeed
Warning

A174371-100G A174371-25G A174371-5G
76974-168EA 43 USD
76974-168 76974-170 76974-172 76974-174
1,3,5-Tris(4-(tert-butyl)-3-hydroxy-2,6-dimethylbenzyl)-1,3,5-triazinane-2,4,6-trione 97%
1,3,5-Tris(4-(tert-butyl)-3-hydroxy-2,6-dimethylbenzyl)-1,3,5-triazinane-2,4,6-trione

Precaución: for Research Use Only

Formula: C₄₂H₅₇N₃O₆
MDL Number: MFCD00192555
CAS Number: 40601-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR