Ethyl-3-amino-4-(cyclohexylamino)benzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ferrostatin-1, Ethyl 3-amino-4-(cyclohexylamino)benzoate
A228648-10MG
A228648-1MG
A228648-25MG
A228648-50MG
A228648-5MG
A228648-100MG
A228648-1G
A228648-250MG
A228648-5G
76976-048EA
36.08
USD
76976-048
76976-050
76976-052
76976-054
76976-056
77600-826
77600-828
77612-216
77672-026
Ethyl-3-amino-4-(cyclohexylamino)benzoate 98%
Ethyl-3-amino-4-(cyclohexylamino)benzoate
Precaución: for Research Use Only
Formula:
C₁₅H₂₂N₂O₂ MW: 262.35 g/mol |
MDL Number:
MFCD08072959 CAS Number: 347174-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...