4,4'-Bis(2-methoxystyryl)biphenyl 97%

Supplier: Ambeed
Warning

A625360-25G A625360-500G A625360-5G
76980-650EA 8 USD
76980-650 76980-652 76980-654
4,4'-Bis(2-methoxystyryl)biphenyl 97%
4,4'-Bis(2-methoxystyryl)biphenyl

Precaución: for Research Use Only

Formula: C₃₀H₂₆O₂
MDL Number: MFCD23160777
CAS Number: 40470-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR